Learn More
Medchemexpress LLC L-phenylalanine, 3-[[[2,4-dicyano-3-[4-[2-(methylamino)-2-oxoethoxy]phenyl]pyrido[1,2-a]benzimidazol-1-yl)amino]c... | 1941174-13-5 | 99.8% | 100 MG
SDP
Catalog No. 5000231829
Encompass_Preferred
KMH-233 is a potent, reversible, and selective inhibitor of the L-type amino acid transporter LAT1 (SLC7A5), used in research to block uptake of neutral amino acids and to study LAT1-mediated nutrient transport and cancer cell metabolism.
  • Potent, reversible inhibitor of LAT1 (IC50 ≈ 18 μM).
  • Reduces cancer cell growth (MCF-7 cell growth IC50 ≈ 124 μM).
  • Enhances anticancer drug efficacy in combination studies.
  • Molecular formula C32H25N7O5; molecular weight 587.58.
  • High purity (99.75% by LCMS).
  • Solid, white to yellow; store powder at -20°C for long-term stability.
Catalog No. 50-002-31829 Supplier Medchemexpress LLC Supplier No. HY120139100MG
KMH-233 is a potent, reversible, and selective inhibitor of the L-type amino acid transporter LAT1 (SLC7A5), used in research to block uptake of neutral amino acids and to study LAT1-mediated nutrient transport and cancer cell metabolism.
  • Potent, reversible inhibitor of LAT1 (IC50 ≈ 18 μM).
  • Reduces cancer cell growth (MCF-7 cell growth IC50 ≈ 124 μM).
  • Enhances anticancer drug efficacy in combination studies.
  • Molecular formula C32H25N7O5; molecular weight 587.58.
  • High purity (99.75% by LCMS).
  • Solid, white to yellow; store powder at -20°C for long-term stability.