Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Phenylalanine, 98.5+%

Catalog No. AC130310025
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130310025 2.5 kg Plastic Bottle
AC130310250 25 g Plastic Bottle
AC130311000 100 g Plastic Bottle
AC130315000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC130310025 Supplier Thermo Scientific Chemicals Supplier No. 130310025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 270.0°C to 275.0°C
pH 5.4 to 6.0 (1% aq. soln.)
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Lead (Pb) 10ppm max.
Loss on Drying 0.3% max. (105°C, 3 hrs)
Assay Percent Range 98.5+%
Packaging Plastic Bottle
Linear Formula C6H5CH2CH(NH2)CO2H
Specific Rotation − 34.10
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol
Specific Rotation Condition − 34.10 (20.00°C c=2,water,dry basis)
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
Quantity 2.5 kg
Formula Weight 165.19
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Phenylalanine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.