Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Phenylalanine, >99%, MP Biomedicals

Catalog No. ICN19472380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19472380 100 g
ICN19472325 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19472380 Supplier MP Biomedicals, Inc Supplier No. 0219472380
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Quantity 100 g
Formula Weight 165.2
Percent Purity 100%
Grade Cell Culture
Chemical Name or Material L-Phenylalanine, >99%
Recommended Storage Room temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.