Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-phenylalanine-carboxy-13C, N-[(9H-fluoren-9-ylmethoxy)carbonyl]- | 916262-84-5 | BD01843532 | 99.8% | 388.42 | C23(13C)H21NO4 | 10 MG
SDP

Catalog No. 5000298383
Encompass_Preferred

Fmoc-Phe-OH-13C is a 13C isotopically labeled, Fmoc-protected L-phenylalanine provided as a powder for research applications including stable-isotope labeling, peptide synthesis, and mass spectrometry tracing.

  • 13C isotopic label with ~99% enrichment.
  • High purity: 99.8% (HPLC).
  • Molecular weight: 388.42 g·mol⁻1.
  • Supplied as a 10 mg powder for laboratory use.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-98383 Supplier Medchemexpress LLC Supplier No. HY79131S510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fmoc-Phe-OH-13C is a 13C isotopically labeled, Fmoc-protected L-phenylalanine provided as a powder for research applications including stable-isotope labeling, peptide synthesis, and mass spectrometry tracing.

  • 13C isotopic label with ~99% enrichment.
  • High purity: 99.8% (HPLC).
  • Molecular weight: 388.42 g·mol⁻1.
  • Supplied as a 10 mg powder for laboratory use.
  • Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.