Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-phenylalanine, glycylglycyl- | 6234-26-0 | MFCD00066054 | 99.9% | 279.30 g/mol | C13H17N3O4 | 1 G
SDP

Catalog No. 5000208786
Encompass_Preferred

H-Gly-Gly-Phe-OH is a tripeptide (glycyl-glycyl-L-phenylalanine) supplied as a solid for research use, commonly used as a cleavable linker motif in antibody-drug conjugate synthesis and peptide chemistry research.

  • Used as a tripeptide linker in ADC and peptide synthesis applications.
  • Tripeptide sequence: Gly-Gly-Phe.
  • CAS number: 6234-26-0.
  • Molecular weight: 279.30 g/mol.
  • Typical physical form: powder; store at 4 °C and protect from light.
  • High-purity material supplied; verify certificate of analysis for exact purity.

Catalog No. 50-002-08786 Supplier Medchemexpress LLC Supplier No. HYW0909401G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-Gly-Gly-Phe-OH is a tripeptide (glycyl-glycyl-L-phenylalanine) supplied as a solid for research use, commonly used as a cleavable linker motif in antibody-drug conjugate synthesis and peptide chemistry research.

  • Used as a tripeptide linker in ADC and peptide synthesis applications.
  • Tripeptide sequence: Gly-Gly-Phe.
  • CAS number: 6234-26-0.
  • Molecular weight: 279.30 g/mol.
  • Typical physical form: powder; store at 4 °C and protect from light.
  • High-purity material supplied; verify certificate of analysis for exact purity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.