Learn More
Medchemexpress LLC L-phenylalanine, L-gamma-glutamyl- | 7432-24-8 | MFCD00002629 | 99.8% | 294.30 g/mol | C14H18N2O5 | 50 MG
SDP
Catalog No. 5000211142
Encompass_Preferred
γ-Glutamylphenylalanine (gamma-Glu-Phe) is a purified dipeptide reagent provided as a solid for research and analytical use. It is reported to enhance umami intensity in food model systems and is commonly used as an endogenous metabolite standard in biochemical and flavor studies. Available in small milligram-scale packages with a high listed purity.
Catalog No. 50-002-11142 Supplier Medchemexpress LLC Supplier No. HY10139950MG
γ-Glutamylphenylalanine (gamma-Glu-Phe) is a purified dipeptide reagent provided as a solid for research and analytical use. It is reported to enhance umami intensity in food model systems and is commonly used as an endogenous metabolite standard in biochemical and flavor studies. Available in small milligram-scale packages with a high listed purity.