Learn More
Medchemexpress LLC L-phenylalanine, L-γ-glutamyl- | 7432-24-8 | 99.8% | 294.30 g/mol | C14H18N2O5 | 10 MG
SDP
Catalog No. 5000213056
Encompass_Preferred
γ-Glu-Phe (gamma-glutamylphenylalanine) is a dipeptide research reagent supplied as a high-purity powder. It is used in flavor chemistry and endogenous metabolism studies and has been reported to enhance umami intensity in food matrices. The material is intended for laboratory research and is accompanied by analytical documentation for quality control.
Catalog No. 50-002-13056 Supplier Medchemexpress LLC Supplier No. HY10139910MG
γ-Glu-Phe (gamma-glutamylphenylalanine) is a dipeptide research reagent supplied as a high-purity powder. It is used in flavor chemistry and endogenous metabolism studies and has been reported to enhance umami intensity in food matrices. The material is intended for laboratory research and is accompanied by analytical documentation for quality control.