Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-phenylalanine, N-(2-pyrazinylcarbonyl)- | 114457-94-2 | MFCD09800536 | 99.7% | 271.28 g/mol | C14H13N3O3 | 25 G
SDP

Catalog No. 5000303872
Encompass_Preferred

Antibacterial agent 268 (N-(Pyrazinylcarbonyl)-L-phenylalanine; CAS 114457-94-2) is a small-molecule antibacterial compound provided as a solid with high reported purity for research applications, including antimycobacterial screening and in vitro assays.

  • Molecular formula: C14H13N3O3.
  • Molecular weight: 271.28 g/mol.
  • Appearance: solid powder.
  • Purity: 99.74% (HPLC) as reported.
  • Suitable for in vitro antibacterial and antimycobacterial studies.
  • Stable under recommended storage conditions for long-term use.

Catalog No. 50-003-03872 Supplier Medchemexpress LLC Supplier No. 25GHYW018811
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antibacterial agent 268 (N-(Pyrazinylcarbonyl)-L-phenylalanine; CAS 114457-94-2) is a small-molecule antibacterial compound provided as a solid with high reported purity for research applications, including antimycobacterial screening and in vitro assays.

  • Molecular formula: C14H13N3O3.
  • Molecular weight: 271.28 g/mol.
  • Appearance: solid powder.
  • Purity: 99.74% (HPLC) as reported.
  • Suitable for in vitro antibacterial and antimycobacterial studies.
  • Stable under recommended storage conditions for long-term use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.