Learn More
Medchemexpress LLC L-phenylalanine, N-[(9H-fluoren-9-ylmethoxy)carbonyl]glycylglycyl-, 1,1-dimethylethyl ester | 236426-37-2 | 99.8% | 557.64 g/mol | C32H35N3O6 | 50 MG
SDP
Catalog No. 5000201952
Encompass_Preferred
Fmoc-Gly-Gly-Phe-OtBu (CAS 236426-37-2) is an Fmoc-protected tripeptide used as a cleavable linker building block in the synthesis of antibody-drug conjugates (ADCs) and as an intermediate for peptide synthesis.
  • Fmoc-protected N-terminus for standard peptide coupling
  • Phenylalanine-containing cleavable linker sequence for ADC construction
  • High purity suitable for research use (99.8%)
  • Available in multiple sizes from 5 mg to 500 mg
Catalog No. 50-002-01952 Supplier Medchemexpress LLC Supplier No. HY4423450MG
Fmoc-Gly-Gly-Phe-OtBu (CAS 236426-37-2) is an Fmoc-protected tripeptide used as a cleavable linker building block in the synthesis of antibody-drug conjugates (ADCs) and as an intermediate for peptide synthesis.
  • Fmoc-protected N-terminus for standard peptide coupling
  • Phenylalanine-containing cleavable linker sequence for ADC construction
  • High purity suitable for research use (99.8%)
  • Available in multiple sizes from 5 mg to 500 mg