Learn More
Cambridge Isotope Laboratories L-Phenylalanine (Ring-D5) | 56253-90-8 | MFCD00144664 | 1 g
Catalog No. NC1136934
Encompass
L-Phenylalanine (Ring-D5) | Purity: 98% | Mol Wt: 170.22 | 56253-90-8 | MFCD00144664 | 1 g
Catalog No. NC1136934 Supplier Cambridge Isotope Laboratories Supplier No. DLM12581
L-Phenylalanine (Ring-D5) | Purity: 98% | Mol Wt: 170.22 | 56253-90-8 | MFCD00144664 | 1 g