Learn More
Bioworld L-PHENYLALANINE SEPAROPORE
Catalog No. 50197849
Encompass
L-PHENYLALANINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: L-Phenylalanine.; Particle size (range): 55-175m.; Molecular Weight range: 6x104 - 2x107.; Matrix: Separopore 4B-CL.; pH stability: 3-10.; Flow specifications: 70-140 cm...
Catalog No. 50-197-849 Supplier Bioworld Supplier No. 201810711
L-PHENYLALANINE SEPAROPORE (AGAROSE) 4B-CL: Specifications: Ligand: L-Phenylalanine.; Particle size (range): 55-175m.; Molecular Weight range: 6x104 - 2x107.; Matrix: Separopore 4B-CL.; pH stability: 3-10.; Flow specifications: 70-140 cm...