Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Phenylalanine (White Crystals or Crystalline Powder), Fisher BioReagents

Catalog No. BP391100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP391-100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP391-100

missing translation for 'validMainframeMsg'

This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
IUPAC Name 2-amino-3-phenylpropanoic acid
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 270°C
Ammonium 0.02% max.
Arsenic (As) 1ppm max.
pH 5.4 to 6.0
Color White
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Assay Percent Range 98.5 to 100.5 %
Loss on Drying 0.20% max.
Other Amino Acids Not Detected
Packaging Glass Bottle
Residue after Ignition 0.1% max.
Specific Rotation -33 to -35° (+ or -)
Merck Index 15, 7382
State of Solution (%T) 98.0% min.
Alpha Vector Phenylalanine
Quantity 100 g
Formula Weight 165.19
Percent Purity 98.5 to 100.5%
Physical Form Powder
Chemical Name or Material L-Phenylalanine
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.