Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Phenylalaninol, 98%

Catalog No. p-7081565
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1158606 5 g
AAA1158614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1158606 Supplier Thermo Scientific Chemicals Supplier No. A1158606
Only null left
Add to Cart

CAS: 3182-95-4 | C9H13NO | 151.21 g/mol

L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

Solubility
Soluble in chloroform, ethyl acetate, ethanol and methanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3182-95-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00004732
InChI Key STVVMTBJNDTZBF-VIFPVBQESA-N
Synonym l-phenylalaninol, s-2-amino-3-phenyl-1-propanol, s---2-amino-3-phenyl-1-propanol, s-2-amino-3-phenylpropan-1-ol, h-phenylalaninol, h-phe-ol, 2s-2-amino-3-phenylpropan-1-ol, phenylalaninol, --l-phenylalaninol, l-phenyl alaninol
PubChem CID 447213
IUPAC Name (2S)-2-amino-3-phenylpropan-1-ol
SMILES N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 89°C to 94°C
Quantity 5 g
Optical Rotation −23° (c=5 in ethanol)
Sensitivity Air Sensitive
UN Number UN3259
Beilstein 2208238
Solubility Information Soluble in chloroform,ethyl acetate,ethanol and methanol.
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material L-Phenylalaninol
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
EINECSNumber 221-674-4
RTECSNumber UA6900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.