Learn More
Bachem H-Ser(PO3H2)-OH
SDP
Supplier:  Bachem 4002875.0025
Encompass_Preferred
H-Ser(PO3H2)-OH, 407-41-0, C3H8NO6P, Mr 185.07, 25g. Dexfosfoserine. L-Phosphoserine binds to b-amyloid in vitro, probably to the charged EVHHQK region. The elevated levels of phosphoserine observed in brains of Alzheimer patients could be a protective response.
Catalog No. 50-194-6649