Learn More
Medchemexpress LLC l-Pindolol | 26328-11-0 | MFCD00153863 | 99.9% | 248.32 g/mol | C14H20N2O2 | 1 ML
SDP
Catalog No. 5000214831
Encompass_Preferred
l-Pindolol is the levorotatory isomer of pindolol, provided as a 10 mM solution in a 1 mL vial for research use. It functions as a reversible, competitive 5-HT1A/1B antagonist and a partial β-adrenoceptor agonist, and is used in pharmacology and receptor characterization studies. Purity is listed as greater than 98%, and SDS and datasheet documentation are available.
Catalog No. 50-002-14831 Supplier Medchemexpress LLC Supplier No. HY10060610MM 1ML
l-Pindolol is the levorotatory isomer of pindolol, provided as a 10 mM solution in a 1 mL vial for research use. It functions as a reversible, competitive 5-HT1A/1B antagonist and a partial β-adrenoceptor agonist, and is used in pharmacology and receptor characterization studies. Purity is listed as greater than 98%, and SDS and datasheet documentation are available.