Learn More
Cayman Chemical L-Pipecolic Acid, Purity: ≥95%, CAS: 3105-95-1, 1 g
SDP
Catalog No. 502911754
Encompass_Preferred
A metabolite of L-lysine; inhibits GABA binding in bovine brain homogenate in the presence, but not absence, of hexobarbital or pentobarbital (IC50s = 0.2 and 2 nM, respectively); serum levels are elevated in patients with Zellweger syndrome
Catalog No. 50-291-1754 Supplier Cayman Chemical Supplier No. 363131 G
A metabolite of L-lysine; inhibits GABA binding in bovine brain homogenate in the presence, but not absence, of hexobarbital or pentobarbital (IC50s = 0.2 and 2 nM, respectively); serum levels are elevated in patients with Zellweger syndrome