Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Pipecolic Acid, > 99%, MP Biomedicals™

Catalog No. ICN15189825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
1 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15189825 25 mg
ICN15189880 100 mg
ICN15189883 250 mg
ICN15189891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN15189825 Supplier MP Biomedicals, Inc Supplier No. 0215189825
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3105-95-1
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00005981
InChI Key HXEACLLIILLPRG-UHFFFAOYNA-N
Synonym l-pipecolic acid, s-piperidine-2-carboxylic acid, l-homoproline, h-hopro-oh, l-pipecolinic acid, l-pipecolate, 2s-piperidine-2-carboxylic acid, s-pipecolic acid, l--pipecolinic acid, l-piperidine-2-carboxylic acid
PubChem CID 439227
ChEBI CHEBI:30913
SMILES OC(=O)C1CCCCN1

Specifications

Color White
Assay Percent Range >99%
Quantity 25 mg
IUPAC Name piperidine-2-carboxylic acid
Formula Weight 129.2
Physical Form Solid
Chemical Name or Material L-Pipecolic Acid
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.