Learn More
Medchemexpress LLC L-prolinamide, 3-methyl-L-valyl-4-hydroxy-N-[[4-(4-methyl-5-thiazolyl)phenyl]methyl] hydrochloride | 1448189-80-7 | MFCD30502992 | 99.7% | 467.02 g/mol | C22H31ClN4O3S | 100 MG
SDP
Catalog No. 5000246888
Encompass_Preferred
(S,R,S)-AHPC monohydrochloride is a VH032-based von Hippel-Lindau (VHL) E3 ligase ligand supplied as the hydrochloride salt. It is provided as a high-purity solid intended for use as a PROTAC building block to recruit VHL for targeted protein degradation studies and can be conjugated to target-binding ligands via linkers.
  • High purity suitable for chemical biology applications.
  • Supplied as a stable hydrochloride salt solid.
  • Used as a VHL E3 ligase ligand in PROTAC design and E3 ligase studies.
  • Compatible with common storage conditions (powder at -20°C; in solution at -80°C).
  • Batch certificate of analysis available for quality control.
Catalog No. 50-002-46888 Supplier Medchemexpress LLC Supplier No. HY101763A100MG
(S,R,S)-AHPC monohydrochloride is a VH032-based von Hippel-Lindau (VHL) E3 ligase ligand supplied as the hydrochloride salt. It is provided as a high-purity solid intended for use as a PROTAC building block to recruit VHL for targeted protein degradation studies and can be conjugated to target-binding ligands via linkers.
  • High purity suitable for chemical biology applications.
  • Supplied as a stable hydrochloride salt solid.
  • Used as a VHL E3 ligase ligand in PROTAC design and E3 ligase studies.
  • Compatible with common storage conditions (powder at -20°C; in solution at -80°C).
  • Batch certificate of analysis available for quality control.