Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-prolinamide, D-phenylalanyl-N-[(1S)-4-[(aminoiminomethyl)amino]-1-(chloroacetyl)butyl] TFA | 157379-44-7 | 99.3% | 25 MG
SDP

Catalog No. 5000232164
Encompass_Preferred

PPACK (TFA) is the trifluoroacetate salt of a peptide-based irreversible thrombin inhibitor supplied as a white to off-white solid. It is used in biochemical assays to inhibit thrombin activity and for anticoagulation research.

  • Irreversible thrombin inhibitor for biochemical assays.
  • High purity of 99.3% as determined by HPLC.
  • White to off-white solid form suitable for storage and handling.
  • Recommended storage conditions for powder and solvent are provided in the COA.
  • Molecular weight 564.99 and formula C23H32ClF3N6O5.

Catalog No. 50-002-32164 Supplier Medchemexpress LLC Supplier No. HY122542B25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PPACK (TFA) is the trifluoroacetate salt of a peptide-based irreversible thrombin inhibitor supplied as a white to off-white solid. It is used in biochemical assays to inhibit thrombin activity and for anticoagulation research.

  • Irreversible thrombin inhibitor for biochemical assays.
  • High purity of 99.3% as determined by HPLC.
  • White to off-white solid form suitable for storage and handling.
  • Recommended storage conditions for powder and solvent are provided in the COA.
  • Molecular weight 564.99 and formula C23H32ClF3N6O5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.