Learn More
Medchemexpress LLC L-proline-13C5,15N | 202407-23-6 | 99.6% | 121.09 | 13C5H915NO2 | 1mg
SDP
Catalog No. 5000267536
Encompass_Preferred
L-Proline-13C5 1-15N is the 13C- and 15N-labeled L-Proline L-Proline is one of the twenty amino acids used in living organisms as the building blocks of proteins
Catalog No. 50-002-67536 Supplier Medchemexpress LLC Supplier No. HYY0252S41MG
L-Proline-13C5 1-15N is the 13C- and 15N-labeled L-Proline L-Proline is one of the twenty amino acids used in living organisms as the building blocks of proteins