Learn More
Sigma Aldrich Fine Chemicals Biosciences L-PROLINE, 1KG
Catalog No. NC2209572
Encompass
Synonym(s):(S)-Pyrrolidine-2-carboxylic acid.Cas# 147-85-3from non-animal source, meets EP, USP testing specifications, suitable for cell culture
This item is not returnable. View return policy
Catalog No. NC2209572 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P56071KG
Synonym(s):(S)-Pyrrolidine-2-carboxylic acid.Cas# 147-85-3from non-animal source, meets EP, USP testing specifications, suitable for cell culture