Learn More
Chem-Impex International, Inc. L-Proline-2-chlorotrityl resin (0.3 - 0.8 meq/g, 100 - 200 mesh) | MFCD00801415 | 25G
Catalog No. NC1593618
Encompass_Preferred
L-Proline-2-chlorotrityl resin (0.3 - 0.8 meq/g, 100 - 200 mesh), MFCD00801415, 25G
Catalog No. NC1593618 Supplier Chem-Impex International, Inc. Supplier No. 0348225G
L-Proline-2-chlorotrityl resin (0.3 - 0.8 meq/g, 100 - 200 mesh), MFCD00801415, 25G