Learn More
MilliporeSigma™ L-Proline, Calbiochem™,
Catalog No. 5370100GM Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
53-701-00GM 100 g
1 options
Catalog No. 53-701-00GM Supplier MilliporeSigma™ Supplier No. 5370100GM
Add to Cart
Add to Cart
Non-essential amino acid that serves as a precursor to hydroxyproline in collagen and features heat-stable solutions suitable for autoclaving.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Form Solid
Format Glass Bottle
Quantity 100 g