Learn More
Medchemexpress LLC L-Proline, L-methionyl-L-α-glutamyl-L-histidyl-L-phenylalanyl-L-prolylglycyl-, acetate | 2828433-33-4 | 99.4% | 873.97 | 100 MG
SDP
Catalog No. 5000424796
Encompass_Preferred
Semax acetate is a BBB-penetrable adrenocorticotropic hormone-like peptide that forms stable complexes with Cu2+. It is a synthetic peptide analog of adrenocorticotropic hormone (ACTH) (4-10) and exhibits immunomodulatory, nootropic, and neuroprotective activities. It is used in research for central nervous system diseases such as Alzheimer's disease and cerebral ischemia.
  • Reduces Aβ oligomer levels.
  • Increases survival and stimulates choline acetyltransferase activity in cholinergic neurons.
  • Increases BDNF and NGF levels in glial cells.
  • Ameliorates effects in a rat model of focal cerebral ischemia.
  • Increases 5-HIAA concentration in hypothalamus and striatum of mice.
  • Affects gene expression related to immune and vascular systems in ischemia rats.
Catalog No. 50-004-24796 Supplier Medchemexpress LLC Supplier No. HYP1146A100MG
Semax acetate is a BBB-penetrable adrenocorticotropic hormone-like peptide that forms stable complexes with Cu2+. It is a synthetic peptide analog of adrenocorticotropic hormone (ACTH) (4-10) and exhibits immunomodulatory, nootropic, and neuroprotective activities. It is used in research for central nervous system diseases such as Alzheimer's disease and cerebral ischemia.
  • Reduces Aβ oligomer levels.
  • Increases survival and stimulates choline acetyltransferase activity in cholinergic neurons.
  • Increases BDNF and NGF levels in glial cells.
  • Ameliorates effects in a rat model of focal cerebral ischemia.
  • Increases 5-HIAA concentration in hypothalamus and striatum of mice.
  • Affects gene expression related to immune and vascular systems in ischemia rats.