Learn More
Medchemexpress LLC L-proline, L-threonyl-L-lysyl-L-prolyl-L-arginyl-L-prolylglycyl (diacetate) | 00-00-0 | 99.9% | 871.98 | C37H65N11O13 | 10 MG
SDP
Catalog No. 5000243873
Encompass_Preferred
Selank diacetate is a synthetic peptide derived from tuftsin with reported anxiolytic, nootropic, antidepressant, and antistress activity. It is supplied as a white to off-white solid for laboratory research use only, with documented molecular and purity data to support experimental use. Use appropriate laboratory safety practices and consult the SDS and COA prior to handling.
  • High purity suitable for research applications.
  • Known amino acid sequence: Thr-Lys-Pro-Arg-Pro-Gly-Pro.
  • Supplied as a white to off-white solid.
  • High solubility in water with sonication recommended.
  • COA and SDS available for quality and safety information.
Catalog No. 50-002-43873 Supplier Medchemexpress LLC Supplier No. HY105042A10MG
Selank diacetate is a synthetic peptide derived from tuftsin with reported anxiolytic, nootropic, antidepressant, and antistress activity. It is supplied as a white to off-white solid for laboratory research use only, with documented molecular and purity data to support experimental use. Use appropriate laboratory safety practices and consult the SDS and COA prior to handling.
  • High purity suitable for research applications.
  • Known amino acid sequence: Thr-Lys-Pro-Arg-Pro-Gly-Pro.
  • Supplied as a white to off-white solid.
  • High solubility in water with sonication recommended.
  • COA and SDS available for quality and safety information.