Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Proline Methyl Ester Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. P03425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03425G 5 g
P034225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P03425G Supplier TCI America Supplier No. P03425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2133-40-6 | C6H12NO2 | 130.17 g/mol

Chemical Identifiers

CAS 2133-40-6
Molecular Formula C6H12NO2
Molecular Weight (g/mol) 130.17
MDL Number MFCD00012708
InChI Key BLWYXBNNBYXPPL-YFKPBYRVSA-O
Synonym l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733200
IUPAC Name (2S)-2-(methoxycarbonyl)pyrrolidin-1-ium
SMILES COC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 73°C
Boiling Point 55°C
Color White
Quantity 5 g
Formula Weight 165.62
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material L-Proline Methyl Ester Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.