Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-proline, N-acetyl-L-seryl-L-α-aspartyl-L-lysyl (acetate) | 00-00-0 | MFCD00076849 | 99.1% | 547.56 | C22H37N5O11 | 5 MG
SDP

Catalog No. 5000263012
Encompass_Preferred

Goralatide acetate (Ac-SDKP) is a naturally occurring N-acetylated tetrapeptide used in research as a physiological regulator of hematopoiesis. It inhibits primitive hematopoietic cell proliferation and is supplied as a white to off-white solid with well-documented analytical data for laboratory applications.

  • Physiological regulator of hematopoiesis.
  • Inhibits primitive hematopoietic cell proliferation.
  • High purity (>99.1% by HPLC) suitable for research use.
  • Well-characterized chemical properties: C22H37N5O11; 547.56 Da.
  • White to off-white solid form for convenient handling.
  • Stable when sealed and kept away from moisture; follow recommended cold storage.
  • Available in milligram-scale quantities for laboratory studies.

Catalog No. 50-002-63012 Supplier Medchemexpress LLC Supplier No. HY129283B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goralatide acetate (Ac-SDKP) is a naturally occurring N-acetylated tetrapeptide used in research as a physiological regulator of hematopoiesis. It inhibits primitive hematopoietic cell proliferation and is supplied as a white to off-white solid with well-documented analytical data for laboratory applications.

  • Physiological regulator of hematopoiesis.
  • Inhibits primitive hematopoietic cell proliferation.
  • High purity (>99.1% by HPLC) suitable for research use.
  • Well-characterized chemical properties: C22H37N5O11; 547.56 Da.
  • White to off-white solid form for convenient handling.
  • Stable when sealed and kept away from moisture; follow recommended cold storage.
  • Available in milligram-scale quantities for laboratory studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.