Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Proline, N-acetyl-L-seryl-L-α-aspartyl-L-lysyl- (acetate) | 99.1% | 25 MG
SDP

Catalog No. 5000338786
Encompass_Preferred

Goralatide, a natural tetrapeptide, is a physiological regulator of hematopoiesis and can inhibit primitive haematopoietic cell proliferation.

  • Purity of 99.1%
  • Molecular weight of 547.56
  • Molecular formula C22H37N5O11
  • Appears as a white to off-white solid
  • Sequence shortening: Ac-SDKP
  • Solubility in DMSO: 100 mg/mL (182.63 mM)
  • Physiological regulator of hematopoiesis
  • Inhibits primitive haematopoietic cell proliferation

Catalog No. 50-003-38786 Supplier Medchemexpress LLC Supplier No. 25MGHY129283B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Goralatide, a natural tetrapeptide, is a physiological regulator of hematopoiesis and can inhibit primitive haematopoietic cell proliferation.

  • Purity of 99.1%
  • Molecular weight of 547.56
  • Molecular formula C22H37N5O11
  • Appears as a white to off-white solid
  • Sequence shortening: Ac-SDKP
  • Solubility in DMSO: 100 mg/mL (182.63 mM)
  • Physiological regulator of hematopoiesis
  • Inhibits primitive haematopoietic cell proliferation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.