Learn More
Medchemexpress LLC L-Proline p-nitroanilide TFA | 108321-19-3 | 99.7% | 349.26 | 5 G
SDP
Catalog No. 5000452098
Encompass_Preferred
L-Proline p-nitroanilide (P-pNA) TFA is a proline derivative. This product is a solid that appears white to light yellow.
  • Ships at room temperature in the continental US.
  • Store at 4°C in sealed containers, away from moisture.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, sealed and away from moisture.
Catalog No. 50-004-52098 Supplier Medchemexpress LLC Supplier No. HYW0111555G
L-Proline p-nitroanilide (P-pNA) TFA is a proline derivative. This product is a solid that appears white to light yellow.
  • Ships at room temperature in the continental US.
  • Store at 4°C in sealed containers, away from moisture.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, sealed and away from moisture.