Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Prolinol, MP Biomedicals™

Catalog No. ICN15638201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15638201 1 mL
ICN15638205 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15638201 Supplier MP Biomedicals, Inc Supplier No. 0215638201
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23356-96-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
InChI Key HVVNJUAVDAZWCB-YFKPBYRVSA-N
Synonym l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
PubChem CID 640091
IUPAC Name [(2S)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Quantity 1 mL
Formula Weight 101.1
Physical Form Liquid
Chemical Name or Material L-Prolinol
EINECSNumber 245-605-2
Recommended Storage Store at 2°-8°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.