Learn More
Medchemexpress LLC L-Propargylglycine | 23235-01-0 | 113.11 | 1 G
SDP
Catalog No. 5000448615
Encompass_Preferred
L-Propargylglycine, also known as (S)-2-Aminopent-4-ynoic acid, is a synthetic amino acid. It is utilized in the creation of folate-conjugates and their associated metal-chelate complexes. This compound functions as a click chemistry reagent, containing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Synthetic amino acid
  • Used in the synthesis of folate-conjugates and metal-chelate complexes
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition with Azide groups
  • For research use only
Catalog No. 50-004-48615 Supplier Medchemexpress LLC Supplier No. HYW0060641G
L-Propargylglycine, also known as (S)-2-Aminopent-4-ynoic acid, is a synthetic amino acid. It is utilized in the creation of folate-conjugates and their associated metal-chelate complexes. This compound functions as a click chemistry reagent, containing an Alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules possessing Azide groups.
  • Synthetic amino acid
  • Used in the synthesis of folate-conjugates and metal-chelate complexes
  • Functions as a click chemistry reagent
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition with Azide groups
  • For research use only