Learn More
Medchemexpress LLC L-Propargylglycine | 23235-01-0 | MFCD00077855 | 113.11 | 25 G
SDP
Catalog No. 5000448219
Encompass_Preferred
L-Propargylglycine, also known as (S)-2-Aminopent-4-ynoic acid, is a synthetic amino acid. It is primarily used in the synthesis of folate-conjugates and corresponding metal-chelate complexes. As a click chemistry reagent, it contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Synthetic amino acid
  • Used in synthesis of folate-conjugates
  • Used in synthesis of metal-chelate complexes
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-004-48219 Supplier Medchemexpress LLC Supplier No. 25GHYW006064
L-Propargylglycine, also known as (S)-2-Aminopent-4-ynoic acid, is a synthetic amino acid. It is primarily used in the synthesis of folate-conjugates and corresponding metal-chelate complexes. As a click chemistry reagent, it contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that contain Azide groups.
  • Synthetic amino acid
  • Used in synthesis of folate-conjugates
  • Used in synthesis of metal-chelate complexes
  • Click chemistry reagent
  • Contains an alkyne group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc)