Learn More
Medchemexpress LLC L-Pyroglutamic acid | 98-79-3 | 98.6% | C5H7NO3 | 25 MG
SDP
Catalog No. 5000358410
Encompass_Preferred
L-Pyroglutamic acid is the levo-isomer of Pyroglutamic acid, serving as the biologically active enantiomer in humans. It functions as an intermediate in glutathione metabolism and can be utilized as a biomarker for systemic lupus erythematosus (SLE).
  • Biologically active levo-isomer
  • Intermediate in glutathione metabolism
  • Potential biomarker for systemic lupus erythematosus (SLE)
  • Solid appearance, ranging from white to light yellow
  • Classified under ketones, aldehydes, and acids
  • Originates as an endogenous metabolite and disease marker
Catalog No. 50-003-58410 Supplier Medchemexpress LLC Supplier No. 25MGHY76082R
L-Pyroglutamic acid is the levo-isomer of Pyroglutamic acid, serving as the biologically active enantiomer in humans. It functions as an intermediate in glutathione metabolism and can be utilized as a biomarker for systemic lupus erythematosus (SLE).
  • Biologically active levo-isomer
  • Intermediate in glutathione metabolism
  • Potential biomarker for systemic lupus erythematosus (SLE)
  • Solid appearance, ranging from white to light yellow
  • Classified under ketones, aldehydes, and acids
  • Originates as an endogenous metabolite and disease marker