Learn More
Cayman Chemical L-Pyrohomoglutamic Acid, Purity: ≥95%, CAS: 34622-39-4, 1 g
SDP
Catalog No. 502988034
Encompass_Preferred
An amino acid building block; has been used in the synthesis of ligands for FKBPs and HDAC inhibitors
Catalog No. 50-298-8034 Supplier Cayman Chemical Supplier No. 316941 G
An amino acid building block; has been used in the synthesis of ligands for FKBPs and HDAC inhibitors