Learn More
Medchemexpress LLC L-pyrohomoglutamic acid | 34622-39-4 | MFCD03839863 | 97.0% | 143.14 g·mol⁻¹ | C6H9NO3 | 5 G
SDP
Catalog No. 5000301450
Encompass_Preferred
L-pyrohomoglutamic acid is a non-proteinogenic amino acid used as a synthetic building block in medicinal chemistry and biochemical research. It is employed as an intermediate for preparing ligands targeting FK506-binding proteins (FKBPs) and histone deacetylase (HDAC) inhibitors, and is supplied as a purified powder for laboratory use.
  • High purity (~97%) suitable for research applications.
  • Used as a building block for FKBP and HDAC ligand synthesis.
  • Molecular formula C6H9NO3 and molecular weight 143.14 g·mol⁻¹.
  • Available in lab-scale pack sizes for synthetic and analytical workflows.
  • Provided with product documentation including COA and SDS for quality assurance.
Catalog No. 50-003-01450 Supplier Medchemexpress LLC Supplier No. HYW0175425G
L-pyrohomoglutamic acid is a non-proteinogenic amino acid used as a synthetic building block in medicinal chemistry and biochemical research. It is employed as an intermediate for preparing ligands targeting FK506-binding proteins (FKBPs) and histone deacetylase (HDAC) inhibitors, and is supplied as a purified powder for laboratory use.
  • High purity (~97%) suitable for research applications.
  • Used as a building block for FKBP and HDAC ligand synthesis.
  • Molecular formula C6H9NO3 and molecular weight 143.14 g·mol⁻¹.
  • Available in lab-scale pack sizes for synthetic and analytical workflows.
  • Provided with product documentation including COA and SDS for quality assurance.