Learn More
Medchemexpress LLC L-pyrohomoglutamic acid | 34622-39-4 | MFCD03839863 | 97.9% | 143.14 | C6H9NO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01754210G
Encompass_Preferred
L-Pyrohomoglutamic acid is an amino acid-derived building block used in organic synthesis and medicinal chemistry. It is employed as a precursor for ligands that bind FK506-binding proteins and as an intermediate in the synthesis of histone deacetylase inhibitor scaffolds. The product is supplied for research use with documented purity and supporting documentation.
  • Amino acid-derived building block for synthetic chemistry.
  • Used to prepare FK506-binding protein ligands and HDAC inhibitor scaffolds.
  • Molecular formula C6H9NO3; molecular weight 143.14.
  • Available in multiple pack sizes, including 10 g.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-002-99446