Learn More
Medchemexpress LLC L-pyrohomoglutamic acid | 34622-39-4 | MFCD03839863 | 97.9% | 143.14 g·mol⁻¹ | C6H9NO3 | 25 G
SDP
Catalog No. 5000303103
Encompass_Preferred
L-pyrohomoglutamic acid is an amino-acid building block used in small-molecule synthesis. It serves as a precursor in medicinal chemistry for preparing ligands that target FK506-binding proteins (FKBPs) and histone deacetylase (HDAC) inhibitors, and is supplied for research and drug-intermediate applications.
  • Amino-acid building block for small-molecule synthesis.
  • Precursor for FKBP and HDAC ligand synthesis.
  • High purity (~97.9%) suitable for research applications.
  • Molecular formula C6H9NO3; molecular weight 143.14 g·mol⁻¹.
  • Available in multiple pack sizes, including 25 g.
  • Includes COA and SDS documentation for quality and safety.
Catalog No. 50-003-03103 Supplier Medchemexpress LLC Supplier No. 25GHYW017542
L-pyrohomoglutamic acid is an amino-acid building block used in small-molecule synthesis. It serves as a precursor in medicinal chemistry for preparing ligands that target FK506-binding proteins (FKBPs) and histone deacetylase (HDAC) inhibitors, and is supplied for research and drug-intermediate applications.
  • Amino-acid building block for small-molecule synthesis.
  • Precursor for FKBP and HDAC ligand synthesis.
  • High purity (~97.9%) suitable for research applications.
  • Molecular formula C6H9NO3; molecular weight 143.14 g·mol⁻¹.
  • Available in multiple pack sizes, including 25 g.
  • Includes COA and SDS documentation for quality and safety.