Learn More
Cayman Chemical L-Pyrohomoglutamic Acid, Purity: ≥95%, CAS: 34622-39-4, 500 mg
SDP
Catalog No. 502992289
Encompass_Preferred
An amino acid building block; has been used in the synthesis of ligands for FKBPs and HDAC inhibitors
Catalog No. 50-299-2289 Supplier Cayman Chemical Supplier No. 31694500 MG
An amino acid building block; has been used in the synthesis of ligands for FKBPs and HDAC inhibitors