Learn More
Medchemexpress LLC HY-N2375 5mg Medchemexpress, L-Quebrachitol CAS:642-38-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN23755MG
Encompass_Preferred
Medchemexpress, HY-N2375 5mg L-Quebrachitol CAS:642-38-6 L-Quebrachitol is a natural product isolated from many plants, promotes osteoblastogenesis by uppregulation of BMP-2, runt-related transcription factor-2 (Runx2), MAPK (ERK, JNK, p38α), and Wnt/β-catenin signaling pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1501
 Restricted Item