Learn More
Cayman Chemical L-QuIsqualIc AcId 10mg
SDP
Catalog No. 502984131
Encompass_Preferred
A natural analog of glutamate that acts as an agonist at AMPA-selective and metabotropic glutamate receptors (EC50s = 170, 10, 40, and 29 nM at GluR-A, mGluR1, mGluR3, and mGluR5, respectively), as well as (GRIK4 (Ki = 6.43 nM)
Catalog No. 50-298-4131 Supplier Cayman Chemical Supplier No. 2021110 MG
A natural analog of glutamate that acts as an agonist at AMPA-selective and metabotropic glutamate receptors (EC50s = 170, 10, 40, and 29 nM at GluR-A, mGluR1, mGluR3, and mGluR5, respectively), as well as (GRIK4 (Ki = 6.43 nM)