Learn More
Medchemexpress LLC L-Rhamnose monohydrate | 10030-85-0 | 182.17 | 1 ML
SDP
Catalog No. 5000415977
Encompass_Preferred
Rhamnose monohydrate is an orally active deoxysugar. It inhibits levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs) in skin aging models. It also stimulates PKA signaling, acts against obesity by stimulating fat dopamine receptors and inducing thermogenesis, and shows anti-aging effects. It can be used in the study of Ehrlich's solid tumors and sarcomas.
  • Orally active deoxysugar
  • Inhibits pro-inflammatory interleukin and MMPs in skin aging models
  • Stimulates PKA signaling by promoting phosphorylation of PKA substrates and HSL in SVF-derived adipocytes
  • Acts against obesity in mice through stimulation of adipose dopamine receptors and inducing thermogenesis
  • Shows anti-aging effects
  • Used in the study of Ehrlich's solid tumors and sarcomas
  • Antitumor activity in mice with Ehrlich's solid tumor
Catalog No. 50-004-15977 Supplier Medchemexpress LLC Supplier No. HYN1420A10MM/1ML
Rhamnose monohydrate is an orally active deoxysugar. It inhibits levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs) in skin aging models. It also stimulates PKA signaling, acts against obesity by stimulating fat dopamine receptors and inducing thermogenesis, and shows anti-aging effects. It can be used in the study of Ehrlich's solid tumors and sarcomas.
  • Orally active deoxysugar
  • Inhibits pro-inflammatory interleukin and MMPs in skin aging models
  • Stimulates PKA signaling by promoting phosphorylation of PKA substrates and HSL in SVF-derived adipocytes
  • Acts against obesity in mice through stimulation of adipose dopamine receptors and inducing thermogenesis
  • Shows anti-aging effects
  • Used in the study of Ehrlich's solid tumors and sarcomas
  • Antitumor activity in mice with Ehrlich's solid tumor