Learn More
Medchemexpress LLC α-L-Rhamnose monohydrate | 6155-35-7 | 182.17 | 500 G
SDP
Catalog No. 5000388685
Encompass_Preferred
α-L-Rhamnose monohydrate is a component of the plant cell wall pectic polysaccharides rhamnogalacturonan I and rhamnogalacturonan II. It is also a component of bacterial polysaccharides where it plays an important role in pathogenicity.
  • Purity: 98.0%
  • For research use only
Catalog No. 50-003-88685 Supplier Medchemexpress LLC Supplier No. HYN0642500G
α-L-Rhamnose monohydrate is a component of the plant cell wall pectic polysaccharides rhamnogalacturonan I and rhamnogalacturonan II. It is also a component of bacterial polysaccharides where it plays an important role in pathogenicity.
  • Purity: 98.0%
  • For research use only