Learn More
Medchemexpress LLC L-Ribose | 24259-59-4 | 150.13 | 100 G
SDP
Catalog No. 5000447440
Encompass_Preferred
L-Ribose is a non-naturally occurring pentose that serves as an ideal starting material for synthesizing L-nucleosides analogues. It can be utilized in the synthesis of various anticancer and antiviral agents.
  • Non-naturally occurring pentose
  • Ideal starting material for synthesizing L-nucleosides analogues
  • Can be used in the synthesis of anticancer and antiviral agents
Catalog No. 50-004-47440 Supplier Medchemexpress LLC Supplier No. HYW008351100G
L-Ribose is a non-naturally occurring pentose that serves as an ideal starting material for synthesizing L-nucleosides analogues. It can be utilized in the synthesis of various anticancer and antiviral agents.
  • Non-naturally occurring pentose
  • Ideal starting material for synthesizing L-nucleosides analogues
  • Can be used in the synthesis of anticancer and antiviral agents