Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-Ribose | 24259-59-4 | 150.13 | 5 G
SDP

Catalog No. 5000446883
Encompass_Preferred

L-Ribose is a non-naturally occurring pentose. It serves as an ideal starting material for synthesizing L-nucleosides analogues. It can be utilized in the synthesis of various anticancer and antiviral agents.

  • Ideal starting material for synthesizing L-nucleosides analogues
  • Utilized in the synthesis of various anticancer and antiviral agents
  • Purity: 98.0%
  • Molecular weight: 150.13
  • Formula: C5H10O5
  • Store at -20°C, stored under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (stored under nitrogen)

Catalog No. 50-004-46883 Supplier Medchemexpress LLC Supplier No. HYW0083515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ribose is a non-naturally occurring pentose. It serves as an ideal starting material for synthesizing L-nucleosides analogues. It can be utilized in the synthesis of various anticancer and antiviral agents.

  • Ideal starting material for synthesizing L-nucleosides analogues
  • Utilized in the synthesis of various anticancer and antiviral agents
  • Purity: 98.0%
  • Molecular weight: 150.13
  • Formula: C5H10O5
  • Store at -20°C, stored under nitrogen
  • In solvent, store at -80°C for 6 months or -20°C for 1 month (stored under nitrogen)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.