Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ribulose | 2042-27-5 | >96.0% | 10 MG
SDP

Catalog No. 5000233824
Encompass_Preferred

L-ribulose is an isomer of L-ribose that serves as a precursor for the synthesis of rare sugars and L-nucleoside analogues. It is supplied as a research-grade chemical reagent in small laboratory pack sizes and is intended for research use only (not for human use).

  • Isomer of L-ribose used as a precursor in rare sugar synthesis.
  • High purity (≥96.0%).
  • Available in small research packs, including 10 mg.
  • Molecular formula C5H10O5; molecular weight 150.13 g·mol-1.
  • Recommended storage: -20°C for long-term stability; in solvent store at -80°C for extended periods.
  • Intended for research use only, not for clinical or human use.

Catalog No. 50-002-33824 Supplier Medchemexpress LLC Supplier No. HY121305A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ribulose is an isomer of L-ribose that serves as a precursor for the synthesis of rare sugars and L-nucleoside analogues. It is supplied as a research-grade chemical reagent in small laboratory pack sizes and is intended for research use only (not for human use).

  • Isomer of L-ribose used as a precursor in rare sugar synthesis.
  • High purity (≥96.0%).
  • Available in small research packs, including 10 mg.
  • Molecular formula C5H10O5; molecular weight 150.13 g·mol-1.
  • Recommended storage: -20°C for long-term stability; in solvent store at -80°C for extended periods.
  • Intended for research use only, not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.