Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ribulose | 2042-27-5 | 99.9% | 1 MG
SDP

Catalog No. 5000232876
Encompass_Preferred

L-ribulose is an isomer of L-ribose provided as a 1 M aqueous solution for research use; it serves as a precursor for synthesis of rare sugars and L-nucleoside analogues.

  • 1 M concentration in water.
  • CAS number 2042-27-5.
  • Purity 99.9% (as supplied).
  • Molecular formula C5H10O5; molecular weight 150.13 g/mol.
  • Supplied in micro-quantities suitable for synthesis and analytical work.
  • Intended for research use only.

Catalog No. 50-002-32876 Supplier Medchemexpress LLC Supplier No. HY121305B1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-ribulose is an isomer of L-ribose provided as a 1 M aqueous solution for research use; it serves as a precursor for synthesis of rare sugars and L-nucleoside analogues.

  • 1 M concentration in water.
  • CAS number 2042-27-5.
  • Purity 99.9% (as supplied).
  • Molecular formula C5H10O5; molecular weight 150.13 g/mol.
  • Supplied in micro-quantities suitable for synthesis and analytical work.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.