Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ribulose | 2042-27-5 | 99.9% | 10 MG
SDP

Catalog No. 5000232363
Encompass_Preferred

L-Ribulose (solution) is supplied as a 1 M aqueous solution of L-ribulose for research use. L-ribulose is an isomer of L-ribose and is commonly used as a precursor in the synthesis of rare sugars and L-nucleoside analogues.

  • Contains 1 M L-ribulose in water, equivalent to 10 mg of solute.
  • Reported purity of 99.85% (manufacturer listing).
  • Appearance: liquid, colorless to light yellow.
  • Intended for research use; suitable as a building block for carbohydrate and nucleoside synthesis.

Catalog No. 50-002-32363 Supplier Medchemexpress LLC Supplier No. HY121305B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ribulose (solution) is supplied as a 1 M aqueous solution of L-ribulose for research use. L-ribulose is an isomer of L-ribose and is commonly used as a precursor in the synthesis of rare sugars and L-nucleoside analogues.

  • Contains 1 M L-ribulose in water, equivalent to 10 mg of solute.
  • Reported purity of 99.85% (manufacturer listing).
  • Appearance: liquid, colorless to light yellow.
  • Intended for research use; suitable as a building block for carbohydrate and nucleoside synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.