Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC L-ribulose | 2042-27-5 | 99.9% | 5 MG
SDP

Catalog No. 5000234850
Encompass_Preferred

L-Ribulose (solution) is a 1 M aqueous solution of L-ribulose (CAS 2042-27-5) supplied as a 5 mg pre-dosed solution. It is used as a precursor for the synthesis of rare sugars and L-nucleoside analogue research applications.

  • 1 M concentration of L-ribulose in water.
  • Contains 5 MG of L-ribulose per vial (1 M · 33.3 μL).
  • Molecular formula C5H10O5, molecular weight 150.13 g·mol⁻1.
  • High purity (approximately 99.85%).
  • Provided as a small-volume, pre-dosed solution for convenience in analytical and synthetic workflows.

Catalog No. 50-002-34850 Supplier Medchemexpress LLC Supplier No. HY121305B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Ribulose (solution) is a 1 M aqueous solution of L-ribulose (CAS 2042-27-5) supplied as a 5 mg pre-dosed solution. It is used as a precursor for the synthesis of rare sugars and L-nucleoside analogue research applications.

  • 1 M concentration of L-ribulose in water.
  • Contains 5 MG of L-ribulose per vial (1 M · 33.3 μL).
  • Molecular formula C5H10O5, molecular weight 150.13 g·mol⁻1.
  • High purity (approximately 99.85%).
  • Provided as a small-volume, pre-dosed solution for convenience in analytical and synthetic workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.