Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Precision Pump Tubing, PharmaPure, L/S 16; 25 ft
Catalog No. NC9442917
Encompass
Applications: Pharmaceutical and biotech Characteristics: Long life under pressure up to 60 psi (4 bar), excellent biocompatibility, low extractables, very low spallation, nontoxic, nonhemolytic, opaque off-white
Catalog No. NC9442917 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX0643516/DEL
Applications: Pharmaceutical and biotech Characteristics: Long life under pressure up to 60 psi (4 bar), excellent biocompatibility, low extractables, very low spallation, nontoxic, nonhemolytic, opaque off-white