Learn More
Med Vet International Zoetis L-S 50 Water Soluble Powder, 75gm
Catalog No. 502828869
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Lincomycin-spectinomycin (antibacterial and antimycoplasmal) for use in chickens up to seven days of age as an aid in the control of Airsacculitis caused by either Mycoplasma synoviae or Mycoplasma gallisepticum susceptible to lincomycin-spectinomycin, and Complicated Chronic Respiratory Disease (Air Sac Infection) caused by Escherichia coli and M. gallisepticum susceptible to lincomycin-spectinomycin. One bottle treats 25 gallons of drinking water.
Catalog No. 50-282-8869 Supplier Med Vet International Supplier No. RXLINSPEC75
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Lincomycin-spectinomycin (antibacterial and antimycoplasmal) for use in chickens up to seven days of age as an aid in the control of Airsacculitis caused by either Mycoplasma synoviae or Mycoplasma gallisepticum susceptible to lincomycin-spectinomycin, and Complicated Chronic Respiratory Disease (Air Sac Infection) caused by Escherichia coli and M. gallisepticum susceptible to lincomycin-spectinomycin. One bottle treats 25 gallons of drinking water.